Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136491.1 | NP_001129963.1 | 3 - 4 | 490 | 68 | |
GenBank mRNA | FJ211194.1 | - | 3 - 4 | 490 | 68 |
Gene Symbol: LOC114118377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979143.1 | XP_027834944.1 | 3 - 4 | 551 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |