Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161880.1 | NP_001155352.1 | 1 - 2 | 434 | 61 | |
XM_012100421.3 | XP_011955811.1 | 3 - 4 | 668 | 61 | ||
XM_012100423.3 | XP_011955813.1 | 2 - 3 | 554 | 61 | ||
XM_015102518.2 | XP_014958004.1 | 3 - 4 | 673 | 61 | ||
GenBank mRNA | FJ943992.1 | - | 1 - 2 | 434 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Endogenous Controls Recommended Assay Ovis aries |