Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161886.1 | NP_001155358.1 | 4 - 5 | 704 | 101 | |
XM_012110487.3 | XP_011965877.1 | 4 - 5 | 942 | 101 | ||
GenBank mRNA | FJ958367.1 | - | 4 - 5 | 704 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |