Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162546.1 | NP_001156018.1 | 5 - 6 | 447 | 101 | |
GenBank mRNA | FJ969400.1 | - | 5 - 6 | 447 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |