Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009418.2 | NP_001009418.1 | 2 - 3 | 188 | 99 | |
GenBank mRNA | S53077.1 | - | 2 - 3 | 189 | 99 | |
Z12969.1 | - | 189 | 99 |