Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163056.1 | NP_001156528.1 | 4 - 5 | 849 | 83 | |
GenBank mRNA | FJ958356.1 | - | 4 - 5 | 849 | 83 |