Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257093.1 | NP_001244022.1 | 5 - 6 | 945 | 90 | |
XM_012096672.3 | XP_011952062.2 | 5 - 6 | 1356 | 90 | ||
GenBank mRNA | HM245791.1 | - | 3 - 4 | 501 | 90 | |
JQ348894.1 | - | 5 - 6 | 945 | 90 | ||
KF833357.1 | - | 5 - 6 | 1044 | 90 | ||
M63123.1 | - | 5 - 6 | 924 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |