Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009382.1 | NP_001009382.1 | 4 - 5 | 440 | 81 | |
GenBank mRNA | X52993.1 | - | 440 | 81 |