Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009798.1 | NP_001009798.1 | 2 - 3 | 213 | 88 | |
GenBank mRNA | X15493.1 | - | 213 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |