Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126352.1 | NP_001119824.1 | 1 | 759 | 75 | |
XM_015095469.2 | XP_014950955.1 | 8 - 9 | 872 | 75 | ||
XM_015095470.2 | XP_014950956.1 | 8 - 9 | 853 | 75 | ||
XM_027969158.1 | XP_027824959.1 | 8 - 9 | 845 | 75 | ||
GenBank mRNA | EU596596.1 | - | 1 | 759 | 75 |
Gene Symbol: LOC114116592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973508.1 | XP_027829309.1 | 1 | 784 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |