Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126361.1 | NP_001119833.1 | 2 - 3 | 163 | 100 | |
XM_012174025.2 | XP_012029415.1 | 3 - 4 | 329 | 100 | ||
XM_012174619.3 | XP_012030009.1 | 3 - 4 | 346 | 100 | ||
GenBank mRNA | EU596600.1 | - | 2 - 3 | 163 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |