Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126361.1 | NP_001119833.1 | 3 - 4 | 241 | 81 | |
XM_012174025.2 | XP_012029415.1 | 4 - 5 | 407 | 81 | ||
XM_012174619.3 | XP_012030009.1 | 4 - 5 | 424 | 81 | ||
GenBank mRNA | EU596600.1 | - | 3 - 4 | 241 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |