Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260478.1 | NP_001247407.1 | 2 - 3 | 263 | 82 | |
GenBank mRNA | HE617662.1 | - | 2 - 3 | 263 | 82 |