Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009420.2 | NP_001009420.1 | 1 - 2 | 199 | 107 | |
GenBank mRNA | Z18291.1 | - | 199 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |