Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162560.1 | NP_001156032.1 | 2 - 3 | 211 | 97 | |
GenBank mRNA | FJ969404.1 | - | 2 - 3 | 211 | 97 |