Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009276.1 | NP_001009276.1 | 6 - 7 | 858 | 101 | |
XM_027963886.1 | XP_027819687.1 | 6 - 7 | 1008 | 101 | ||
GenBank mRNA | AY208678.1 | - | 6 - 7 | 858 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |