Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009400.2 | NP_001009400.1 | 4 - 5 | 710 | 113 | |
GenBank mRNA | DQ152996.1 | - | 2 - 3 | 208 | 113 | |
X76916.1 | - | 710 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |