Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHLHE40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129741.1 | NP_001123213.1 | 4 - 5 | 385 | 93 | |
GenBank mRNA | EF494246.1 | - | 4 - 5 | 385 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |