Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162562.1 | NP_001156034.1 | 1 - 2 | 163 | 103 | |
XM_015092738.2 | XP_014948224.1 | 4 - 5 | 789 | 103 | ||
XM_027963866.1 | XP_027819667.1 | 2 - 3 | 457 | 103 | ||
GenBank mRNA | FJ958354.1 | - | 1 - 2 | 163 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |