Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162564.1 | NP_001156036.1 | 7 - 8 | 1056 | 121 | |
GenBank mRNA | FJ958360.1 | - | 7 - 8 | 1056 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |