Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104931.1 | NP_001098401.1 | 3 - 4 | 758 | 82 | |
XM_027977360.1 | XP_027833161.1 | 3 - 4 | 855 | 82 | ||
GenBank mRNA | EU072419.1 | - | 3 - 4 | 758 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |