Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104931.1 | NP_001098401.1 | 4 - 5 | 897 | 86 | |
XM_027977360.1 | XP_027833161.1 | 4 - 5 | 994 | 86 | ||
GenBank mRNA | EU072419.1 | - | 4 - 5 | 897 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |