Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST3GAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112818.1 | NP_001106289.1 | 2 - 3 | 115 | 110 | |
XM_012101525.3 | XP_011956915.1 | 3 - 4 | 254 | 110 | ||
GenBank mRNA | AM888395.1 | - | 2 - 3 | 115 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |