Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127269.1 | NP_001120741.1 | 2 - 3 | 358 | 98 | |
XM_012158548.3 | XP_012013938.2 | 3 - 4 | 577 | 98 | ||
GenBank mRNA | EU525168.1 | - | 2 - 3 | 244 | 98 | |
EU626223.1 | - | 2 - 3 | 358 | 98 | ||
FJ943997.1 | - | 2 - 3 | 358 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |