Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127285.1 | NP_001120757.1 | 2 - 3 | 237 | 84 | |
XM_012097249.3 | XP_011952639.1 | 5 - 6 | 7224 | 84 | ||
XM_012097252.3 | XP_011952642.1 | 4 - 5 | 7188 | 84 | ||
XM_012097256.3 | XP_011952646.1 | 5 - 6 | 6976 | 84 | ||
XM_012097257.3 | XP_011952647.1 | 5 - 6 | 7336 | 84 | ||
XM_012097260.3 | XP_011952650.1 | 7 - 8 | 3801 | 84 | ||
XM_027980221.1 | XP_027836022.1 | 5 - 6 | 5519 | 84 | ||
XM_027980222.1 | XP_027836023.1 | 5 - 6 | 4362 | 84 | ||
XM_027980223.1 | XP_027836024.1 | 6 - 7 | 3885 | 84 | ||
GenBank mRNA | EU626204.1 | - | 2 - 3 | 237 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |