Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130935.1 | NP_001124407.1 | 1 - 2 | 165 | 54 | |
XM_012106097.2 | XP_011961487.1 | 2 - 3 | 205 | 54 | ||
XM_012106098.3 | XP_011961488.1 | 2 - 3 | 264 | 54 | ||
XM_012106099.3 | XP_011961489.1 | 2 - 3 | 269 | 54 | ||
XM_012106101.3 | XP_011961491.1 | 2 - 3 | 208 | 54 | ||
XM_012106102.3 | XP_011961492.1 | 2 - 3 | 282 | 54 | ||
XM_027962744.1 | XP_027818545.1 | 2 - 3 | 269 | 54 | ||
XM_027962745.1 | XP_027818546.1 | 2 - 3 | 282 | 54 | ||
GenBank mRNA | EU797609.1 | - | 1 - 2 | 165 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |