Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134307.1 | NP_001127779.1 | 2 - 3 | 226 | 96 | |
XM_027962474.1 | XP_027818275.1 | 3 - 4 | 450 | 96 | ||
XM_027962475.1 | XP_027818276.1 | 3 - 4 | 373 | 96 | ||
GenBank mRNA | EU797612.1 | - | 2 - 3 | 226 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |