Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009460.1 | NP_001009460.1 | 4 - 5 | 893 | 70 | |
XM_027977373.1 | XP_027833174.1 | 5 - 6 | 2415 | 70 | ||
GenBank mRNA | U14128.1 | - | 4 - 5 | 893 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |