Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009731.1 | NP_001009731.1 | 6 - 7 | 1447 | 60 | |
XM_027969188.1 | XP_027824989.1 | 5 - 6 | 2300 | 60 | ||
GenBank mRNA | AF110984.1 | - | 6 - 7 | 1447 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |