Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009796.1 | NP_001009796.1 | 4 - 5 | 913 | 168 | |
XM_027975567.1 | XP_027831368.1 | 3 - 4 | 870 | 168 | ||
GenBank mRNA | X03883.1 | - | 913 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |