Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139451.1 | NP_001132923.1 | 3 - 4 | 692 | 79 | |
XM_015093768.2 | XP_014949254.1 | 3 - 4 | 719 | 79 | ||
XM_015093770.2 | XP_014949256.1 | 3 - 4 | 704 | 79 | ||
GenBank mRNA | EU293837.1 | - | 3 - 4 | 692 | 79 | |
HM055899.1 | - | 3 - 4 | 638 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |