Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161891.1 | NP_001155363.1 | 6 - 7 | 909 | 99 | |
XM_027975553.1 | XP_027831354.1 | 5 - 6 | 774 | 99 | ||
GenBank mRNA | FJ969421.1 | - | 6 - 7 | 909 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |