Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174111.1 | NP_001167582.1 | 1 - 2 | 46 | 58 | |
GenBank mRNA | FJ715737.1 | - | 1 - 2 | 46 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries Inventoried |