Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318075.1 | NP_001305004.1 | 1 | 2621 | 111 | |
XM_012184941.3 | XP_012040331.1 | 3 | 2878 | 111 | ||
XM_012184942.2 | XP_012040332.1 | 3 | 2882 | 111 | ||
XM_012184943.3 | XP_012040333.1 | 3 | 3006 | 111 | ||
XM_012184944.3 | XP_012040334.1 | 2 | 4791 | 111 | ||
XM_012184946.2 | XP_012040336.1 | 2 | 2706 | 111 | ||
XM_012184947.3 | XP_012040337.1 | 2 | 2831 | 111 | ||
GenBank mRNA | KT180155.1 | - | 1 | 2621 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |