Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040290.1 | NP_001035380.1 | 3 - 4 | 376 | 65 | |
XM_012177596.2 | XP_012032986.2 | 6 - 7 | 2107 | 65 | ||
XM_012177597.3 | XP_012032987.2 | 6 - 7 | 1814 | 65 | ||
XM_027969200.1 | XP_027825001.1 | 5 - 6 | 2268 | 65 | ||
XM_027969201.1 | XP_027825002.1 | 5 - 6 | 2208 | 65 | ||
XM_027969202.1 | XP_027825003.1 | 5 - 6 | 2239 | 65 | ||
XM_027969203.1 | XP_027825004.1 | 5 - 6 | 2268 | 65 | ||
XM_027969206.1 | XP_027825007.1 | 5 - 6 | 2162 | 65 | ||
XM_027969207.1 | XP_027825008.1 | 5 - 6 | 671 | 65 | ||
XM_027969208.1 | XP_027825009.1 | 6 - 7 | 1799 | 65 | ||
GenBank mRNA | DQ130017.1 | - | 3 - 4 | 376 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |