Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131030.1 | NP_001124502.1 | 4 - 5 | 535 | 78 | |
GenBank mRNA | EU797604.1 | - | 4 - 5 | 535 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |