Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163603.1 | NP_001157075.1 | 5 - 6 | 427 | 97 | |
GenBank mRNA | FJ937948.1 | - | 5 - 6 | 427 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |