Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009469.1 | NP_001009469.1 | 6 - 7 | 844 | 79 | |
XM_027973955.1 | XP_027829756.1 | 7 - 8 | 958 | 79 | ||
XM_027973956.1 | XP_027829757.1 | 7 - 8 | 1043 | 79 | ||
XM_027973957.1 | XP_027829758.1 | 7 - 8 | 884 | 79 | ||
GenBank mRNA | X80024.1 | - | 844 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |