Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC66A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126356.1 | NP_001119828.1 | 2 - 3 | 332 | 69 | |
XM_027960679.1 | XP_027816480.1 | 3 - 4 | 483 | 69 | ||
GenBank mRNA | EU583473.1 | - | 2 - 3 | 332 | 69 |