Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPARC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166202.1 | NP_001159674.1 | 3 - 4 | 209 | 71 | |
XM_012177565.3 | XP_012032955.1 | 4 - 5 | 389 | 71 | ||
GenBank mRNA | GQ221054.1 | - | 3 - 4 | 209 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |