Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPARC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166202.1 | NP_001159674.1 | 7 - 8 | 732 | 176 | |
XM_012177565.3 | XP_012032955.1 | 8 - 9 | 912 | 176 | ||
GenBank mRNA | GQ221054.1 | - | 7 - 8 | 732 | 176 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |