Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174110.1 | NP_001167581.1 | 1 | 624 | 118 | |
GenBank mRNA | FJ751853.1 | - | 1 | 624 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |