Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280691.1 | NP_001267620.1 | 4 - 5 | 1028 | 93 | |
GenBank mRNA | KC857456.1 | - | 4 - 5 | 1028 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |