Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040289.1 | NP_001035379.1 | 4 - 5 | 488 | 87 | |
GenBank mRNA | DQ458790.1 | - | 4 - 5 | 488 | 87 |