Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078660.1 | NP_001072128.1 | 4 - 5 | 347 | 73 | |
XM_027977374.1 | XP_027833175.1 | 5 - 6 | 493 | 73 | ||
XM_027977375.1 | XP_027833176.1 | 5 - 6 | 519 | 73 | ||
GenBank mRNA | DQ496115.1 | - | 4 - 5 | 347 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |