Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168634.1 | NP_001162105.1 | 6 - 7 | 1051 | 82 | |
GenBank mRNA | FN568103.1 | - | 6 - 7 | 1051 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |