Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256817.1 | NP_001243746.1 | 3 - 4 | 372 | 83 | |
GenBank mRNA | HE652877.1 | - | 3 - 4 | 372 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |