Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009189.1 | NP_001009189.1 | 1 - 2 | 950 | 88 | |
GenBank mRNA | AB018478.1 | - | 1 - 2 | 950 | 88 | |
HQ263386.1 | - | 1 - 2 | 911 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |