Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006603.4 | XP_004006652.1 | 3 - 4 | 446 | 77 | |
XM_012174781.3 | XP_012030171.1 | 3 - 4 | 448 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |