Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962485.1 | XP_027818286.1 | 4 - 5 | 1497 | 109 | |
XM_027962486.1 | XP_027818287.1 | 5 - 6 | 1424 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |